Order Entry
Netherlands
ContactUsLinkComponent
1,5-Diphenyl-3-(p-tolyl)formazan ≥95.0% (by HPLC)
1,5-Diphenyl-3-(p-tolyl)formazan ≥95.0% (by HPLC)
Catalog # TCIAB0338-100MG
Supplier:  TCI
CAS Number:  
1,5-Diphenyl-3-(p-tolyl)formazan ≥95.0% (by HPLC)
Catalog # TCIAB0338-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Red to Dark blue to Black powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Melting point:
    167.0 to 175.0 deg-C
  • Molar extinction coefficient:
    min. 12000 (acetonitrile, 484.0 to 488.0nm)

Specification Test Results

About this item

 :
 :314.39 g/mol
CAS Number:1622-12-4
Formula:C₂₀H₁₈N₄
MDL Number:MFCD00059675