Order Entry
Netherlands
ContactUsLinkComponent
1,5-Diphenyl-3-(2-thienyl)formazan ≥96.0% (by HPLC)
1,5-Diphenyl-3-(2-thienyl)formazan ≥96.0% (by HPLC)
Catalog # TCIAB0326-100MG
Supplier:  TCI
CAS Number:  
1,5-Diphenyl-3-(2-thienyl)formazan ≥96.0% (by HPLC)
Catalog # TCIAB0326-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Black powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Elemental analysis (Nitrogen):
    17.00 to 19.00 %
  • Lambda max.:
    515.0 to 525.0 nm (CH3CN)
  • Molar extinction coefficient:
    min. 13000 (CH3CN, 515.0 to 525.0 nm)

Specification Test Results

About this item

 :
 :306.39 g/mol
CAS Number:135984-01-9
Formula:C₁₇H₁₄N₄S
MDL Number:MFCD00059744