
1-(4-Nitrophenyl)-3-methyl-5-phenylformazan ≥98.0% (by HPLC)
TCIAB0312-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 94.0 %
- Melting point:157.0 to 161.0 deg-C
:
:283.29 g/mol
:1325
:4.1,III
:GHS02
:68420-26-8
:C₁₄H₁₃N₅O₂
:MFCD00059829