Order Entry
Netherlands
ContactUsLinkComponent
3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride ≥98.0% (by HPLC)
 
3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride ≥98.0% (by HPLC)
  TCIAB0311-100MG
 :  TCI
 :  
undefined
 
3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride ≥98.0% (by HPLC)
  TCIAB0311-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    White to Yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 95.0 %

 

 

 :
 :317.73 g/mol
 :1325
 :4.1,III
 :GHS02
 :C₁₄H₁₂ClN₅O₂
 :MFCD00060011