Saccharin sodium dihydrate ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:6155-57-3
:
Formule:C₇H₅NNaO₃S·2H₂O
MDL-nummer:MFCD00151213
:242.21 g/mol
EINECS:204-886-1
Specificatie testresultaten
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:13.0 to 16.0 %
Specificatie testresultaten
Frequently Bought Together