Order Entry
Netherlands
ContactUsLinkComponent
1,4-Benzoquinone dioxime ≥95.0% (by total nitrogen basis)
Irritant
Health
1,4-Benzoquinone dioxime ≥95.0% (by total nitrogen basis)
Catalog # TCIAB0090-25G
Supplier:  TCI
CAS Number:  
Irritant
Health
1,4-Benzoquinone dioxime ≥95.0% (by total nitrogen basis)
Catalog # TCIAB0090-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown to Dark green powder to crystal
  • Purity (with Total Nitrogen):
    min. 95.0 %

Specification Test Results

About this item

 :
 :138.13 g/mol
EINECS:203-271-5
 :GHS07
 :GHS08
CAS Number:105-11-3
Formula:C₆H₆N₂O₂
MDL Number:MFCD00063636