

Nickel(II) benzenesulphonate hexahydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAB0034-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Gray to Dark blue powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
About this item
:
:373.02 g/mol
EINECS:254-642-3
UN:3077
ADR:9,III
:GHS08
:GHS09
CAS Number:39819-65-3
Formula:C₆H₆O₃S
MDL Number:MFCD00070595