

Benzenesulphonic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:98-11-3
:
Formula:C₆H₆O₃S
MDL Number:MFCD00011689
:158.18 g/mol
UN:2585
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Gray to Red powder to lump
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:transparency
- Water:9.0 to 12.0 %
Specification Test Results
Frequently Bought Together