
DBD-F [=4-(N,N-dimethylaminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥98.0% (by HPLC) for HPLC labeling
Catalog # TCIAA5595-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:124.0 to 128.0 deg-C
- Effect test of HPLC derivatization:Effective as labeling agent for L-Valine.
Specification Test Results
About this item
:
:245.23 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:98358-90-8
Formula:C₈H₈FN₃O₃S
MDL Number:MFCD00143356