
4-Bromomethyl-6,7-dimethoxycoumarin ≥98.0% (by HPLC) for HPLC labeling
Supplier: TCI
Certificates
About this item
CAS Number:88404-25-5
:
Formula:C₁₂H₁₁BrO₄
MDL Number:MFCD00011570
:299.12 g/mol
Storage Temperature:Koelkast
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:211.0 to 216.0 deg-C
- Effect test of HPLC derivatization:Effective as labeling agent for Linoleic Acid , Oleic Acid
Specification Test Results
Frequently Bought Together