Order Entry
Netherlands
ContactUsLinkComponent
4-Nitro-1,2-phenylenediamine monohydrochloride ≥99.0% (by titrimetric analysis) sensitive reagent for the determination of Se by GC-ECD
Irritant
Health
4-Nitro-1,2-phenylenediamine monohydrochloride ≥99.0% (by titrimetric analysis) sensitive reagent for the determination of Se by GC-ECD
Catalog # TCIAA5081-5G
Supplier:  TCI
CAS Number:  
Irritant
Health
4-Nitro-1,2-phenylenediamine monohydrochloride ≥99.0% (by titrimetric analysis) sensitive reagent for the determination of Se by GC-ECD
Catalog # TCIAA5081-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 99.0 %
  • Solubility in 1mol/L HCl:
    almost transparency

Specification Test Results

About this item

 :
 :189.60 g/mol
EINECS:258-429-6
 :GHS07
 :GHS08
CAS Number:53209-19-1
Formula:C₆H₇N₃O₂
MDL Number:MFCD00043273