Farnesyl thiosalicylic acid ≥96.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:162520-00-5
:
Formula:C₂₂H₃₀O₂S
MDL Number:MFCD00467723
:358.54 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Neutralization titration):min. 96.0 %
- Melting point:70.0 to 74.0 deg-C
Specification Test Results
Frequently Bought Together