N-(2-Acryloyloxyethyl)-N-benzyl-N,N-dimethylammonium chloride ∼75% in water stabilised
: TCI
:46830-22-2
:
:C₁₄H₂₀ClNO₂
:MFCD04038840
:269.77 g/mol
:Stabilised with 4-methoxyphenol (MEHQ)
:Refrigerator
:256-283-8
- Appearance:Colorless to Light yellow clear liquid
- Purity (HPLC):min. 90.0 area%
- Purity (Argentmetric Titration):74.0 to 77.0 %
- Water:23.0 to 26.0 %
Frequently Bought Together