Order Entry
Netherlands
ContactUsLinkComponent
 
3-[(3-Acrylamidopropyl)dimethylammonio]propanoate ≥95.0% (by HPLC)
 :  TCI
 
3-[(3-Acrylamidopropyl)dimethylammonio]propanoate ≥95.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

 

Frequently Bought Together