Order Entry
Netherlands
ContactUsLinkComponent
Azido-PEG4-C2-carboxylic acid ≥98.0% (by titrimetric analysis)
Azido-PEG4-C2-carboxylic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAA3260-100MG
Supplier:  TCI
CAS Number:  
undefined
Azido-PEG4-C2-carboxylic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAA3260-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

Synonyms: 3-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethoxy]propanoic acid, Azido-PEG4-C2-acid, 1-Azido-3,6,9,12-tetraoxapentadecan-15-oic acid, Azido-PEG4-C2-carboxylic Acid
 :
 :291.30 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:1257063-35-6
Formula:C₁₁H₂₁N₃O₆
MDL Number:MFCD12406155