
m-β-Hydroxyethylsulphonyl aniline ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5246-57-1
:
Formula:C₈H₁₁NO₃S
MDL Number:MFCD00044070
EINECS:226-046-3
:GHS07
Specification Test Results
- Appearance:White to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:73.0 to 77.0 deg-C
Specification Test Results
Frequently Bought Together