
2-Amino-5-bromo-3-(trifluoromethyl)pyridine ≥98.0% (by GC)
Catalog # TCIAA3139-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g/mol
- Seq:9220
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:88.0 to 92.0 deg-C
Specification Test Results
About this item
:
:241.01 g/mol
:GHS07
CAS Number:79456-34-1
Formula:C₆H₄BrF₃N₂
MDL Number:MFCD10758088