Order Entry
Netherlands
ContactUsLinkComponent
Fmoc-L-Lys(N3)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-L-Lys(N3)-OH ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA3023-250MG
Supplier:  TCI
CAS Number:  
undefined
Fmoc-L-Lys(N3)-OH ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA3023-250MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -11.0 to -15.0 deg (C=1, DMF)

Specification Test Results

About this item

 :
 :394.431 g/mol
Storage Temperature:Refrigerator
CAS Number:159610-89-6
Formula:C₂₁H₂₂N₄O₄
MDL Number:MFCD13182319