4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine ≥98.0% (by HPLC)
TCIAA3020-250MG
: TCI
:
- Pk:250 mg
- Seq:2590
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Melting point:124.0 to 128.0 deg-C
- Specific rotation [a]20/D:-27.0 to -31.0 deg (C=1, DMF)
:
:366.38 g/mol
:Refrigerator
:942518-20-9
:C₁₉H₁₈N₄O₄
:MFCD11052920