2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:27776-21-2
:
Formula:C₁₂H₂₄Cl₂N₆
MDL Number:MFCD00142723
:323.27 g/mol
EINECS:248-655-3
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):25.00 to 27.00 %
Specification Test Results
Frequently Bought Together