
Azido-PEG11-amine ≥93.0% (by HPLC)
: TCI
:1800414-71-4
:749244-38-0
:C₂₄H₅₀N₄O₁₁
:570.68 g/mol
:GHS07
:Refrigerator
- Appearance:Colorless to Almost colorless clear liquid
- Purity (HPLC):min. 93.0 area%
- Elemental analysis (Nitrogen):9.00 to 10.50 %
Frequently Bought Together