
3-Amino-4-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:175204-80-5
:
Formula:C₆H₅F₃N₂
MDL Number:MFCD00173960
:162.11 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to lump
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:54.0 to 58.0 deg-C
Specification Test Results
Frequently Bought Together