2'-Azido-2'-deoxyuridine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:26929-65-7
:
Formula:C₉H₁₁N₅O₅
MDL Number:MFCD00043045
Storage Temperature:Refrigerator
EINECS:248-113-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):25.00 to 27.00 %
Specification Test Results
Frequently Bought Together