
2-Acetoxypropionyl chloride ≥97.0% (by GC, titration analysis)
: TCI
:38939-83-2
:
:C₅H₇ClO₃
:MFCD18969061
:150.56 g/mol
:58°C
:Refrigerator
:3265
:8,III
:GHS05
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
Frequently Bought Together