
4-Amino-2-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
TCIAA2861-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
:
:6.1,III
:162.1100 g/mol
:2811
:GHS06
:147149-98-2
:C₆H₅F₃N₂
:MFCD09056877