
2-Amino-9,9-diphenylfluorene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1268519-74-9
:
Formula:C₂₅H₁₉N
MDL Number:MFCD22376664
:333.43 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together