25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane ≥90.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:869718-80-9
:
Formula:C₁₇H₃₅N₃O₈
MDL Number:MFCD13184962
:409.48 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (HPLC):min. 90.0 area%
Specification Test Results
Related Information
Frequently Bought Together