

3-(4-Methylpiperazin-1-yl)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:148546-99-0
:
Formula:C₁₁H₁₇N₃
MDL Number:MFCD08435857
:191.28 g/mol
UN:3263
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:83.0 to 87.0 deg-C
Specification Test Results
Frequently Bought Together