N-Acetyl-DL-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15962-46-6
:
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD00055924
:193.20 g/mol
EINECS:240-098-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:196.0 to 200.0 deg-C
Specification Test Results
Frequently Bought Together