

4-Azido-2,3,5,6-tetrafluorobenzoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA2674-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Yellow powder to crystalline
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:235.10 g/mol
Storage Temperature:Freezer
UN:1325
ADR:4.1,III
:GHS02
:GHS07
CAS Number:122590-77-6
Formula:C₇HF₄N₃O₂
MDL Number:MFCD00467436