Order Entry
Netherlands
ContactUsLinkComponent
4-(1,3-Benzothiazol-2-yl)phenylamine ≥97.0% (by GC, titration analysis)
Irritant
4-(1,3-Benzothiazol-2-yl)phenylamine ≥97.0% (by GC, titration analysis)
Catalog # TCIAA2672-200MG
Supplier:  TCI
CAS Number:  
Irritant
4-(1,3-Benzothiazol-2-yl)phenylamine ≥97.0% (by GC, titration analysis)
Catalog # TCIAA2672-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    154.0 to 158.0 deg-C

Specification Test Results

About this item

 :
 :226.3 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:6278-73-5
Formula:C₁₃H₁₀N₂S
MDL Number:MFCD00182896