Amiloride hydrochloride ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAA2599-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 % (calcd.on anh.substance)
- Water:8.0 to 15.0 %
Specification Test Results
About this item
:
:266.09 g/mol
Melting Pt:299°C
EINECS:217-958-2
CAS Number:2016-88-8
:17440-83-4
Formula:C₆H₈ClN₇O·HCl
MDL Number:MFCD00058197