
Isoquinoline-4-amine ≥98.0% (by GC, titration analysis)
: TCI
:23687-25-4
:
:C₉H₈N₂
:MFCD00034752
:144.1700 g/mol
:245-823-8
:GHS07
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:108.0 to 112.0 deg-C
Frequently Bought Together