trans-4-Aminoadamantan-1-ol hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAA2594-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
Synonyms: trans-1-Amino-4-hydroxyadamantane hydrochloride, trans-4-Amino-1-adamantanol hydrochloride
:
:203.7100 g/mol
:GHS07
CAS Number:62075-23-4
Formula:C₁₀H₁₈ClNO
MDL Number:MFCD11099880