S-(Acetamidomethyl)-N-(tert-butoxycarbonyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA2553-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-38.0 to -41.0 deg (C=1、H2O)
Specification Test Results
About this item
:
:292.35 g/mol
EINECS:243-267-0
CAS Number:19746-37-3
Formula:C₁₁H₂₀N₂O₅S
MDL Number:MFCD00038252