
Adapalene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:106685-40-9
:
Formula:C₂₈H₂₈O₃
MDL Number:MFCD03106112
:412.53 g/mol
Merck Index:14,00150
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together