3-Amino-3-phenyl-1-propanol ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14593-04-5
:
Formula:C₉H₁₃NO
MDL Number:MFCD00801124
Boiling Pt:134°C
Molecular Weight:151.21 g/mol
UN:3259
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:73.0 to 77.0 deg-C
Specification Test Results
Frequently Bought Together