
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥98.0% (by HPLC)
TCIAA2528-50MG
: TCI
:
- Pk:50 mg
- Seq:2370
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):21.20 to 22.00 %
- Specific rotation [a]20/D:-61.0 to -66.0 deg (C=1、H2O)
:
:258.23 g/mol
:14,00016
:220-097-5
:GHS07
:Freezer
:2627-69-2
:C₉H₁₄N₄O₅
:MFCD00869751