Biotin-PEG3-azide ≥95.0% (by HPLC)
TCIAA2523-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Light yellow powder to crystalline
- Purity (HPLC):min. 95.0 area%
- Melting point:107.0 to 112.0 deg-C
:
:444.55 g/mol
:Freezer
:875770-34-6
:C₁₈H₃₂N₆O₅S
:MFCD20134145