L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1713265-25-8
:113170-55-1
Formula:C₁₂H₁₂Mg₃O₁₈P₂·xH₂O
MDL Number:MFCD03428049
:579.07 g/mol (anhy)
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Chelometric Titration):min. 85.0 % (calcd.on anh.substance)
- Water:max. 25.0 %
Specification Test Results
Frequently Bought Together