Order Entry
Netherlands
ContactUsLinkComponent
(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Irritant
(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +27.0 to +30.0 deg (C=0.2, H2O)

Specification Test Results

Related Information

Frequently Bought Together