3-Amino-9H-pyrido[3,4-b]indole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:73834-77-2
:
Formula:C₁₁H₉N₃
MDL Number:MFCD00209818
:183.21 g/mol
Melting Pt:291°C
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together