
2-Azido-1,3-dimethylimidazolinium hexafluorophosphate ≥98.0% (by HPLC)
TCIAA2457-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):23.70 to 24.80 %
:
:285.13 g/mol
:GHS07
:Freezer
:1266134-54-6
:C₅H₁₀F₆N₅P
:MFCD18782891