DL-Alanine methyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:13515-97-4
:
Formula:C₄H₉NO₂
MDL Number:MFCD00035523
:139.58 g/mol
Melting Pt:158°C
EINECS:236-854-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together