
2-Amino-6-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:34486-24-3
:
Formula:C₆H₅F₃N₂
MDL Number:MFCD03093868
:162.11 g/mol
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:85.0 to 89.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together