2,2'-O-Anhydro-5-methyluridine ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAA2431-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:217.0 to 221.0 deg-C
- Specific rotation [a]20/D:-43.0 to -46.0 deg (C=4、H2O)
Specification Test Results
About this item
:
:240.22 g/mol
CAS Number:22423-26-3
Formula:C₁₀H₁₂N₂O₅
MDL Number:MFCD00233555