3-Amino-3-(4-chlorophenyl)propionic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19947-39-8
:
Formula:C₉H₁₀ClNO₂
MDL Number:MFCD00126370
:199.64 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together