
N-(5-Aminopentyl)acetamide ≥97.0% (by GC, titration analysis)
Catalog # TCIAA2393-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:144.22 g/mol
Boiling Pt:110°C
UN:2735
ADR:8,III
:GHS05
CAS Number:32343-73-0
Formula:C₇H₁₆N₂O