
11-Azido-3,6,9-trioxaundecan-1-amine ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:134179-38-7
:
Formula:C₈H₁₈N₄O₃
MDL Number:MFCD00269874
:218.26 g/mol
Storage Temperature:Refrigerator
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 93.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together