
Acamprosate calcium ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAA2359-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:400.47 g/mol
Merck Index:14,00017
Melting Pt:270°C
EINECS:278-665-3
:GHS07
CAS Number:77337-73-6
Formula:C₅H₁₁NO₄S
MDL Number:MFCD00886588